LIF296040627 Life Chemicals Inc.

7-chloro-2-(chloromethyl)quinazolin-4(3H)-one

  • MDL number MFCD04621698
  • Cas Number : 730951-40-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart