LIF296040628 Life Chemicals Inc.

2-(chloromethyl)-9-methyl-4H-pyrido[1,2-a]pyrimidin-4-one

  • MDL number MFCD06370765
  • Cas Number : 796067-44-2
  • Molecular Weight : 208.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart