LIF296040635 Life Chemicals Inc.

7-(chloromethyl)-2-methyl-5H-[1,3]thiazolo[3,2-a]pyrimidin-5-one

  • MDL number MFCD09971918
  • Cas Number : 943656-55-1
  • Molecular Weight : 214.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart