LIF296040649 Life Chemicals Inc.

5-(chloromethyl)-3-(2-thienyl)-1,2,4-oxadiazole

  • MDL number MFCD03407322
  • Cas Number : 63417-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart