LIF296040651 Life Chemicals Inc.

2-(chloromethyl)-5-(4-fluorophenyl)-1,3,4-oxadiazole

  • MDL number MFCD05263166
  • Cas Number : 350672-14-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart