LIF296040653 Life Chemicals Inc.

5-(benzyloxy)-2-(chloromethyl)-4H-pyran-4-one

  • MDL number MFCD04122658
  • Cas Number : 89539-54-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart