LIF372152157 Life Chemicals Inc.

4-(2,5-dimethylphenyl)thiazol-2-amine

  • MDL number MFCD00985673
  • Cas Number : 101967-39-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart