LIF372152160 Life Chemicals Inc.

2-Chloro-7-ethoxy-quinoline-3-carbaldehyde

  • MDL number MFCD00995828
  • Cas Number : 129798-05-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart