LIF372152164 Life Chemicals Inc.

4-(2-naphthyl)-1,3-thiazol-2-amine

  • MDL number MFCD00046452
  • Cas Number : 21331-43-1
  • Molecular Weight : 226.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart