LIF372152171 Life Chemicals Inc.

4-(2-methoxy-1-naphthyl)-1,3-thiazol-2-amine

  • MDL number MFCD01536642
  • Cas Number : 312605-17-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart