LIF372152174 Life Chemicals Inc.

(9H-Purin-6-ylamino)-acetic acid

  • MDL number MFCD00520063
  • Cas Number : 10082-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart