LIF372152175 Life Chemicals Inc.

5-Adamantan-1-yl-2-hydroxy-benzoic acid

  • Cas Number : 126145-51-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart