LIF372152191 Life Chemicals Inc.

[(4-Nitrobenzyl)thio]acetic acid

  • MDL number MFCD00540019
  • Cas Number : 6345-13-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart