LIF372152201 Life Chemicals Inc.

1-(2-Amino-4-phenyl-1,3-thiazol-5-yl)ethanone

  • MDL number MFCD02709943
  • Cas Number : 206555-66-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart