LIF372152205 Life Chemicals Inc.

(1,3-Benzothiazol-2-ylthio)acetic acid

  • MDL number MFCD00227391
  • Cas Number : 6295-57-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart