LIF372152217 Life Chemicals Inc.

2-Thioxo-3-m-tolyl-thiazolidin-4-one

  • MDL number MFCD00087336
  • Cas Number : 23522-38-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart