LIF372152220 Life Chemicals Inc.

2-(2-piperazin-1-ylethyl)-1H-benzo[de]isoquinoline-1,3(2H)-dione

  • MDL number MFCD01166074
  • Cas Number : 66235-11-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart