LIF372152226 Life Chemicals Inc.

4,5-Diphenyl-thiazol-2-ylamine

  • MDL number MFCD00438803
  • Cas Number : 6318-74-7
  • Molecular Weight : 252.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart