LIF372152231 Life Chemicals Inc.

ethyl 5-amino-1H-pyrazole-4-carboxylate

  • MDL number MFCD00005238
  • Cas Number : 6994-25-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart