LIF372152254 Life Chemicals Inc.

3-Benzothiazol-2-yl-4,5,6,7-tetrahydro-benzo[b]thiophen-2-ylamine

  • MDL number MFCD02360332
  • Cas Number : 143361-87-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart