LIF372152255 Life Chemicals Inc.

1-Methyl-1H-benzoimidazole-2-sulfonic acid

  • MDL number MFCD00142670
  • Cas Number : 5533-38-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart