LIF372152256 Life Chemicals Inc.

1-Chloro-2-nitro-4-[(trifluoromethyl)sulfonyl]benzene

  • MDL number MFCD00435125
  • Cas Number : 1550-27-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart