LIF372152269 Life Chemicals Inc.

1-({4-[(Methoxycarbonyl)amino]phenyl}sulfonyl)piperidine-2-carboxylic acid

  • MDL number MFCD00725967
  • Cas Number : 483967-96-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart