LIF372152290 Life Chemicals Inc.

4-Butoxy-N-hydrazinocarbonylmethyl-benzamide

  • MDL number MFCD00700679
  • Cas Number : 392247-14-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart