LIF372152300 Life Chemicals Inc.

(1,1-Dioxido-3-oxo-1,2-benzisothiazol-2(3H)-yl)acetic acid

  • MDL number MFCD00688358
  • Cas Number : 52188-11-1
  • Molecular Weight : 241.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart