LIF372152301 Life Chemicals Inc.

1H-Benzoimidazole-2-sulfonic acid

  • MDL number MFCD00100972
  • Cas Number : 40828-54-4
  • Molecular Weight : 198.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart