LIF372152303 Life Chemicals Inc.

ethyl 2,3-dimethyl-1{H}-indole-5-carboxylate

  • MDL number MFCD00458330
  • Cas Number : 21523-62-6
  • Molecular Weight : 217.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart