LIF372152306 Life Chemicals Inc.

4-Bromo-1-hydroxy-2-naphthoic acid

  • MDL number MFCD00021504
  • Cas Number : 5813-37-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart