LIF372152308 Life Chemicals Inc.

3,6-Dihydroxy-xanthen-9-one

  • MDL number MFCD00226967
  • Cas Number : 1214-24-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart