LIF372152313 Life Chemicals Inc.

3-(4-Bromophenyl)-1-(2-cyanoethyl)-1H-pyrazole-4-carbonitrile

  • MDL number MFCD04407432
  • Cas Number : 475626-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart