LIF372152317 Life Chemicals Inc.

4-[(6-Bromo-4-phenylquinazolin-2-yl)amino]benzoic acid

  • MDL number MFCD02043461
  • Cas Number : 332102-08-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart