LIF372152324 Life Chemicals Inc.

(3-Acetylamino-adamantan-1-yl)-acetic acid

  • Cas Number : 75667-93-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart