LIF372152330 Life Chemicals Inc.

(2E)-3-[4-Methoxy-3-(pyrrolidin-1-ylsulfonyl)phenyl]acrylic acid

  • MDL number MFCD01795563
  • Cas Number : 324779-72-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart