LIF372152351 Life Chemicals Inc.

4-[1-(4-Chloro-phenyl)-1-methyl-ethyl]-phenol

  • MDL number MFCD00626162

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart