LIF372152359 Life Chemicals Inc.

4-(4-Benzylphenyl)-1,3-thiazol-2-amine

  • MDL number MFCD01816130
  • Cas Number : 68729-05-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart