LIF372152361 Life Chemicals Inc.

5-Amino-1-(2-naphthyl)-1H-pyrazole-4-carboxamide

  • MDL number MFCD01912109
  • Cas Number : 307343-86-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart