LIF372152365 Life Chemicals Inc.

N-(Cyanomethyl)-4-methylbenzamide

  • MDL number MFCD00548034
  • Cas Number : 22198-21-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart