LIF372152368 Life Chemicals Inc.

3,5-Dimethyl-4-[(2-nitrophenyl)thio]-1H-pyrazole

  • MDL number MFCD01470728
  • Cas Number : 92289-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart