LIF372152369 Life Chemicals Inc.

(2E)-2-[(4-methylbenzoyl)amino]-3-phenylacrylic acid

  • MDL number MFCD01995434
  • Cas Number : 109448-26-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart