LIF372152378 Life Chemicals Inc.

2,2'-[1,4-Phenylenebis(oxymethylene)]dioxirane

  • MDL number MFCD00186891

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart