LIF372152385 Life Chemicals Inc.

2-(Isobutylthio)-1H-benzimidazole

  • MDL number MFCD00464037
  • Cas Number : 75080-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart