LIF372152392 Life Chemicals Inc.

2-(4-Methoxy-phenyl)-thiazolidine

  • MDL number MFCD00222821
  • Cas Number : 31404-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart