LIF372152403 Life Chemicals Inc.

(5E)-5-(4-hydroxybenzylidene)-2-imino-1,3-thiazolidin-4-one

  • MDL number MFCD00497168
  • Cas Number : 299953-00-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart