LIF372152404 Life Chemicals Inc.

4-(2,6-dimethyl-4-oxopyridin-1(4H)-yl)benzoic acid

  • MDL number MFCD02731053
  • Cas Number : 370585-22-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart