LIF372152405 Life Chemicals Inc.

N'-(4-Hydroxy-6-methylpyrimidin-2-yl)acetohydrazide

  • MDL number MFCD00522220
  • Cas Number : 117098-60-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart