LIF372152406 Life Chemicals Inc.

N1-(4-Bromophenyl)glycinamide

  • MDL number MFCD00790883
  • Cas Number : 325852-86-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart