LIF372152409 Life Chemicals Inc.

methyl 3-hydroxy-5-methylthiophene-2-carboxylate

  • MDL number MFCD01859968
  • Cas Number : 5556-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart