LIF372152411 Life Chemicals Inc.

7-(methylsulfanyl)[1,3]thiazolo[4,5-g][1,3]benzothiazol-2-amine

  • MDL number MFCD03030155
  • Cas Number : 397290-24-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart