LIF372152414 Life Chemicals Inc.

5-Hydroxy-2-phenyl-1-benzofuran-3-carboxylic acid

  • MDL number MFCD01549100
  • Cas Number : 75161-15-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart