LIF372152442 Life Chemicals Inc.

2-(4-methyl-1,3-thiazol-2-yl)acetamide

  • MDL number MFCD02180665
  • Cas Number : 173739-73-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart